Would you like to log in again?
BEAKER PP 250ML GRADUATED PK6
Supplier: Bel-Art Products, a Part of SP
Griffin Low Form Polypropylene Beaker^ 250mL. Graduated. Translucent, chemical resistant and will stack without sticking together. Raised numeric graduations show approximate volume (±5%). Steam autoclavable at 121[degree]C.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BEAKER PP 250ML GRADUATED PK6 47750-440 F262130000 Pack of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty