Would you like to log in again?
SP Bel-Art Conical Tube Holders, 50 ml, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F18795-0000 F18795-0001
47750-481PK 55.56 USD InStock
47750-481 47750-483
SP Bel-Art Conical Tube Holders, 50 ml, Bel-Art Products, a part of SP
Racks Tube Racks
Leave hands free for pipetting and other critical tasks with these polystyrene plastic tube holders that securely hold a 50 mL conical centrifuge tube.
  • Conical bottom design
  • Tubes can be easily removed
  • Available with grip or non-grip style
  • Polystyrene construction
  • Securely fitting hold
Choose from a non-grip style (white) that allows the tube to be lifted freely from holder as needed, or a grip style (blue) that stays with the tube when it is lifted from the bench top. Tubes can easily be removed from grip style holders by holding the base and pulling the tube out.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty