Would you like to log in again?
SP Bel-Art Bel-Blotter™ 96-Well Replicating Tool, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37876-0002
47750-528EA 34.14 USD InStock
47750-528
SP Bel-Art Bel-Blotter™ 96-Well Replicating Tool, Bel-Art Products, a part of SP
Microplate and Petri Dish Replicators
Designed to perform tedious tasks with minimum effort, the 96 open-ended pipet tips of this unique product fit all types of 96 well plates from flat, "V", or round bottom plates to 0.2 mL thin walled PCR plates and tubes.
  • Suited for repetitive routines
  • Polycarbonate construction
  • Easy to use
  • Reusable
  • Autoclavable
The tip will retain up to 10 mL of fluid for placement onto filter paper, plate, or other receiving media.
The tool can be used for replication of recombinant DNA libraries, inoculation of filters for colony hybridization, PCR, phage typing, and other applications. Made of polycarbonate, it is easy-to-use, reusable and autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty