Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art Economy Dry Ice Maker, Bel-Art Products, a part of SP

Supplier: Bel-Art
SP Bel-Art Frigimat®
H38876-0002 H38876-0000
47750-560EA 1286.36 USD InStock
47750-560 47750-562
SP Bel-Art Economy Dry Ice Maker, Bel-Art Products, a part of SP
Dry Ice Makers
Produces a solid 454g (16oz) block of dry ice in one to two minutes.

  • Polyethylene ice chamber
  • Epoxy-coated steel frame
  • Nonskid rubber feet
  • Easy to attach liquid CO2 cylinder
  • Dimension: 37×14×15 cm (14¾×5½×6")
  • Shipping weight: 3.9 kg (8.5 lbs.)

Each 50 lb

The epoxy-coated steel frame has four non-skid rubber feet to prevent sliding. Includes a 1.8m (6ft) nickel-plated copper connecting tube with standard CGA 320 (0.830"-14) thread couplings for easy attachment to a liquid CO2 cylinder.

cylinder of liquid CO2 with a dip tube will make 8 or 9 blocks of dry ice measuring 9.2 x 10.4 cmL (3⅝ x 4"). The polyethylene ice chamber is held in place by a metal clamping mechanism, and a porous polyethylene diffusion plate vents excess gas. Also features a built-in gas pressure gauge and relief valve.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty