Would you like to log in again?
SP Bel-Art Carboys with Spigot, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F11847-0010 F11847-0050 F11847-0020
47750-726EA 176.05 USD InStock
47750-726 47750-728 47750-730
SP Bel-Art Carboys with Spigot, Bel-Art Products, a part of SP
Bottles Carboys
Translucent carboys made of lightweight, durable polyethylene with extra-heavy walls for long life and dependability.
  • Lighter than glass and easy to handle
  • Heavy knurling on cap provides excellent grip for opening and closing
  • Resistant to corrosive acids and alkalis
  • The spigot requires only half turn to open or close
  • Smooth sides accept adhesive labels and can be marked with most indelible markers
Ideal for dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty