Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art Buchner Table-Top Funnels, 457 mm (18"), Bel-Art Products, a part of SP

Supplier: Bel-Art Products, a part of SP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

H14621-0000 H14628-0000 H14626-3518 H14625-3518 H14627-1457 H91462-4457 H14621-1457 H91462-4345 H91463-1457
47750-798EA 1435.2 USD InStock
47750-798 30308-092 47750-788 74950-368 47750-814 74950-372 47750-806 74950-380 13197-328
SP Bel-Art Buchner Table-Top Funnels, 457 mm (18"), Bel-Art Products, a part of SP
Funnels Filtration Funnels Büchner Funnels
Filter large volumes quickly with a polyethylene plastic Table-Top Buchner Funnel using your choice of Fritware® or Perforated filter plate.

  • Removable hose barbed connector accepts 12.7 mm (½") I.D. tube and threads into ¾" NPT bung on the funnel
  • Overall height: 292 mm (11.5")
  • Rim to plate: 203 mm (8")

Fritware® Porous Filter Plates supported by a multiple ring grid, are made of 6.4 mm (¼") thick HDPE, with a non-porous ring at the periphery of the plate which seals filter paper when used. Fritware® Porous Filter Plates (fixed or removable) are available in two porosities (medium, 45 - 90 µm and coarse, 90 - 130 µm). Temperature range: –20...+52 °C (–4...125 °F).

Perforated Filter Plates are made of 4.8 mm (³/₁₆") thick HDPE with 4.8 mm (³/₁₆") perforations on 11 mm (⁷/₁₆") centers. Perforated Filter Plates (fixed or removable): Fixed for coarse filtration or use with a cloth or paper filter; Removable for use in precious metals recovery. Temperature range: –20...+52 °C (–4...125 °F).

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty