Would you like to log in again?
SP Bel-Art Standard Stem Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art
F14676-0055 F14676-0065 F14676-0100 F14676-0075
30617-616CS 224.08 USD InStock
30617-616 30617-618 47750-830 30617-614
SP Bel-Art Standard Stem Funnels, Bel-Art Products, a part of SP
Funnels Filtration Funnels Filter Funnels
Polypropylene plastic funnels feature an exclusive filter-step internal design for speedier filtrations.
  • Polypropylene plastic construction
  • External ribs
  • Autoclavable
  • Filter-step internal design
External ribs prevent airlock. The parallel stem measures 8 mm O.D. Steam autoclavable at 121ºC (250ºF).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty