Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art Vibrasorb® Economy Vibration Damping Mount, Bel-Art Products, a part of SP

Supplier: Bel-Art

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VIBRASORB® SP Bel-Art Vibrasorb®
183850000
47751-010EA 949.82 USD InStock
47751-010
SP Bel-Art Vibrasorb® Economy Vibration Damping Mount, Bel-Art Products, a part of SP
Furniture Tables and Workbenches
Vibrasorb® Vibration Damping Mounts facilitate optimal use and help protect electromechanical and optical equipment from excessive wear and even structural damage that can be caused by exposure to vibration.

  • Protect equipment from excessive wear and structural damage
  • Reduced exposure to vibrations
  • Durable polystyrene shell
  • Smooth, non-slip neoprene working surface

The Vibrasorb® Economy Vibration Damping Mount has a durable, high impact polystyrene shell with a smooth, non-slip neoprene working surface.

Vibro-Absorber™ damping mounts at each corner support the 20.8kg (46 lb) platform. The platform area is 34 x 27.5 cm (13½ x 11") and easily wipes clean. Maximum carrying weight for effective vibration dampening is 11kg (24 lb).

Delivery information: Delivery can be as long as 6-8 weeks.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty