Would you like to log in again?
Sponge Probe Wiper, Advanced Istruments
Supplier: Advanced Instruments
4C3840
47751-466PK 301.49 USD InStock
47751-466
Sponge Probe Wiper, Advanced Istruments
Wipes Sponges
Sponge probe wipers are designed for use with various instruments, specifically for calibration and use with cryoscopes and osmometers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty