Would you like to log in again?
VWR® Polygon Spinbar Magnetic Stirring Bars without Pivot Ring
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
37120-6040 37120-6020 37120-6050 37120-6025 37120-6060 37120-6015 37120-6080 37120-6010 37120-6011 37120-6070 37120-6030
47751-488EA 7.82 USD InStock
47751-488 47751-486 76001-888 47751-484 47751-492 76001-894 47751-482 76001-892 47751-490 47751-480 76001-890
VWR® Polygon Spinbar Magnetic Stirring Bars without Pivot Ring
Stirrers Magnetic Stirring Bars
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Autoclave safe
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty