Would you like to log in again?
SP Bel-Art Connecting Microcentrifuge Tube Racks, Bel-Art, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
189080015 189080005
47751-752EA 41.94 USD InStock
47751-752 47751-750
SP Bel-Art Connecting Microcentrifuge Tube Racks, Bel-Art, Bel-Art Products, a part of SP
Racks Tube Racks
Each of these lightweight racks securely holds 24 microcentrifuge tubes, and individual racks accomodate either 0.5 mL or 1.5–2.0 mL tubes.
  • Durable polycarbonate construction
  • UV-resistant
  • Autoclavable
  • Lightweight design
Each rack features a locking clip that firmly connects racks regardless of tube size, allowing for an expandable, stackable, transportable system that maximizes storage and benchtop space. Racks are constructed from durable polycarbonate, UV-resistant, and steam autoclavable at 121°C (250°F). Racks can be stored at temperatures as low as –80°C.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty