Would you like to log in again?
REGULATOR CGA 590 0 250PSI 1 STAGE BRASS
Supplier: AIRGAS USA, LLC
General-Purpose, Single-Stage Regulator^ Well-suited for closely monitored, short-run non-analytical work. Large, nylon-reinforced, neoprene diaphragm for excellent performance, long life, and easy maintenance. Preset safety relief valve vents to atmosphere.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REGULATOR CGA 590 0 250PSI 1 STAGE BRASS 47752-040 Y11-215F590 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty