Would you like to log in again?
LAMP HOLLOW CATHODE 4 PIN BASE SODIUM
Supplier: BULBTRONICS INC.
HOLLOW CATHODE LAMP^ ELEMENT: SODIUM, DIAMETER: 37MM (1.5inch), DATA CODED FOR VARIAN, WINDOW: UV GLASS ENVELOPE, NEON GAS FILL, MOD0028908, ATOMIC ABSORPTION LAMP
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAMP HOLLOW CATHODE 4 PIN BASE SODIUM 47754-288 0028908 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty