Would you like to log in again?
LAMP FLUORESCENT T8 BLUE 15W 18 IN
Supplier: BULBTRONICS INC.
FLUORESCENT LAMP^ BLE1800B, 15W, SHAPE: T8, OVERALL LENGTH: 18inch, DIAMETER: 25MM, LINEAR, COLOR: BLACKLIGHT BLUE, BASE: G13, MOD0015688 (SPECTRONICS), UVA/LONGWAVE, INSTRUMENTATION LAMP
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAMP FLUORESCENT T8 BLUE 15W 18 IN 47755-488 0015688 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty