Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Starfish Gas/Vacuum Manifold with Connector, Heidolph Radleys
Supplier: Heidolph
15890350
47800-072EA 1281.82 USD InStock
47800-072
Starfish Gas/Vacuum Manifold with Connector, Heidolph Radleys
Manifolds
Gas/Vacuum distribution manifold allows gas or vacuum from a single source to be evenly distributed to up to five positions or vessels (does not control or regulate gas/vacuum flow)
Connectors feature leak-proof shut-off valves.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty