Would you like to log in again?
Richard-Allan Scientific® Cytoseal™ XYL Mounting Medium, Thermo Scientific
Supplier: Thermo Scientific
Cytoseal™
8312-4 8312-4
48212-196CS 584.22 USD InStock
CA48212-196 48212-196
Richard-Allan Scientific® Cytoseal™ XYL Mounting Medium, Thermo Scientific
Stains Microscopy and staining reagents
Inhibit the fading or yellowing of stained specimens with specially formulated Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ XYL mounting media.
  • Xylene-based rapid drying allows microscopic examination of slides soon after application
  • Simple dispensing
This xylene-based mounting medium is made of high-quality synthetic resins and contains an antioxidant to inhibit stain fading and yellowing of specimen. Rapid drying medium allows specimen examination soon after application. Bottle features a twist-open cap for simplified dispensing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty