Would you like to log in again?
Resolve™ Immersion Oil, Epredia™
Supplier: Epredia
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Resolve™
M3004 M4000 M2000 M5000 M3004 M5004 M4004 M2004 M5004 M4004 M4000 M3000 M3000 M5000 M2004 M2000
48218-061CS 211.51 USD InStock
CA-48218-081 CA48218-424 CA48218-402 CA48218-106 48218-061 48218-081 CA48218-049 CA48218-445 CA48218-467 48218-106 48218-424 48218-402 48218-467 48218-445 48218-049 CA48218-061
Resolve™ Immersion Oil, Epredia™
Stains Microscopy and staining reagents
Eliminate toxicity and disposal problems with the Epredia™ Resolve™ immersion oil, which contains no PCBs.
  • Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
  • Packaged conveniently in four sizes
  • Available in both low and high viscosity
  • Non-drying and non-hardening
  • Refractive index of 1.5150
It is a clear, colorless oil that allows light transmission throughout most of the UV-VIS spectrum.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty