Would you like to log in again?
Microscope Immersion Oil, Cargille
Supplier: Cargille
16482-4OZ 16485 16482-16OZ 16490 16252 16482-1/4OZ 16484
470150-324CS 461.6 USD InStock
CA16482 470150-324 48218-504 48218-502 48218-500 48218-510 13984-100
Microscope Immersion Oil, Cargille
Stains Microscopy and staining reagents
Types A and B are virtually interchangeable and are miscible with each other for intermediate viscosities. Produced in larger quantities than other types. Type A, at 150 centistokes, reduces any tendancy to trap air. Type B, at 1250 centistokes, is thick enough for viewing multiple slides with one application.
Types A and B are virtually interchangeable and are miscible with each other for intermediate viscosities
Type NVH is used for inverted, inclined, projection, and long focus instruments. The greater the gap between the cover glass and objective, or between the slide and condenser, the more desirable high viscosity becomes. The very high viscosities of Type NVH at 21,000 cSt give excellent results for these applications.
Type 300 is designed for use with automated hematology systems for successful imaging and mechanical processing. Type 300 is manufactured to meet the stringent requirements of this equipment, which include specialized viscosity and exacting controls for consistency.
Produced in larger quantities than other types. Type A, at 150 centistokes, reduces any tendancy to trap air. Type B, at 1250 centistokes, is thick enough for viewing multiple slides with one application.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty