Would you like to log in again?
LiChroCART® Stainless Steel Superspher® Cartridges, MilliporeSigma
Supplier: MilliporeSigma
LiChroCART® Superspher®
1.16010.0001 1.51909.0001 1.51299.0001 1.16056.0001 1.50973.0001 1.50193.0001 1.16010.1003 1.16010.0001 1.51299.0001 1.50974.0001 1.50193.0001 1.16052.1003 1.50980.0001 1.50792.0001 1.16869.0001 1.16855.0001 1.16857.0001 1.51910.0001 1.51299.1003 1.50791.0001 1.50792.1003 1.16009.0001 1.16052.0001 1.50975.0001 1.16869.0001 1.50198.0001 1.50792.0001 1.50980.0001 1.50975.0001 1.51288.0001 1.51288.0001 1.16051.1003 1.50791.0001 1.16056.1003 1.50198.0001 1.16858.0001 1.50205.0001 1.16051.0001 1.16054.0001 1.16857.0001 1.16855.0001 1.50973.0001 1.16009.0001 1.16054.0001 1.50974.0001 1.16854.0001 1.16051.0001 1.16858.0001 1.16854.0001 1.16052.0001 1.50205.0001
48219-182EA 838.88 USD InStock
48219-182 CA48219-150 48219-184 CA1.50791.0001 CA48219-190 48219-180 CA1.50205.0001 10811-872 CA48219-550 48219-186 10811-870 CA48219-152 10811-948 CA48219-182 CA48219-180 CA1.51288.0001 48219-610 CA48219-186 48219-530 CA48219-184 48219-606 48219-602 10811-836 48219-604 CA48219-610 48219-526 CA48219-170 10143-914 10144-168 EM1.51909.0001 48219-168 CA48219-530 48219-164 48219-166 CA48219-606 48219-916 CA48219-526 CA48219-604 CA48219-602 48219-150 48219-152 48219-190 CA48219-168 CA48219-166 48219-550 10811-982 CA48219-164 10811-980 10811-938 CA48219-916 EM1.50205.0001
LiChroCART® Stainless Steel Superspher® Cartridges, MilliporeSigma
Chromatography Columns
These cartridges are ideal for highly efficient HPLC analysis of complex mixtures where high peak capacity is required, because the guaranteed number of theoretical plates for Superspher® cartridges is <100.000 N/m.
These LiChroCART® cartridges are compatible with manu-CART® '4' parts, which facilitates faster and more flexible method adaptation to smaller bore columns. Endcapped cartridges have a bonded phase that is reacted with trimethylchlorosilane to cover silanols not previously reacted with the initial bonding reagent.
This high performance spherical silica carrier yields an excellent pressure/separation performance ratio according to theoretical calculation and practical experience.
Ordering information: All LiChroCART® cartridges with lengths of 75, 125, or 250 mm require manu-CART® end fittings (14229-736).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty