These cartridges are ideal for highly efficient HPLC analysis of complex mixtures where high peak capacity is required, because the guaranteed number of theoretical plates for Superspher® cartridges is <100.000 N/m.
These LiChroCART® cartridges are compatible with manu-CART® '4' parts, which facilitates faster and more flexible method adaptation to smaller bore columns. Endcapped cartridges have a bonded phase that is reacted with trimethylchlorosilane to cover silanols not previously reacted with the initial bonding reagent.
This high performance spherical silica carrier yields an excellent pressure/separation performance ratio according to theoretical calculation and practical experience.
Ordering information: All LiChroCART® cartridges with lengths of 75, 125, or 250 mm require manu-CART® end fittings (14229-736).