Would you like to log in again?
Purospher® HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
Purospher® LiChroCART®
1.51384.0001 1.50144.0001 1.50169.0001 1.50142.0001 1.50799.0001 1.51384.0001 1.50141.0001 1.50798.0001 1.50144.0001 1.50798.0001 1.50168.0001 1.50167.0001 1.50169.0001 1.50141.0001 1.50167.0001 1.50168.0001 1.50142.0001 1.50799.0001
48219-554EA 1096.4 USD InStock
48219-554 CA48219-520 CA48219-554 48219-520 48219-556 CA48219-310 48219-522 48219-310 48219-518 CA48219-308 48219-308 CA48219-518 CA48219-306 CA48219-522 CA48219-556 48219-306 CA1.51384.0001 10144-170
Purospher® HPLC Columns, MilliporeSigma
Chromatography Columns
Purospher® RP-18 is designed for the separation of problematic basic compounds with simple neutral eluents.
Purospher® RP-18e is a versatile high-purity HPLC column producing excellent peak shapes, designed for both the separation of basic compounds with simple neutral eluents and for the elution of strongly acidic compounds. Excellent separations in a shorter period of time with very good peak symmetry saves time and money. The excellent balance of Purospher® RP-18e in the sum of its chromatographic properties is the key to better separations of complex samples with simpler, neutral eluents. The good chemical stability of Purospher® RP-18e enables the use of mobile phase conditions at pH 8 for a long period of time.
In addition Purospher® RP-18 allows the separation of hydrophilic compounds with aqueous mobile phase. Purospher® RP-18 is based upon a high-purity, metal free silica. The multi-step chemical modification and deactivation of the surface and the polar endcapping enables a peak-tailing free elution of basic and chelating compounds. Purospher® RP-18 is not suitable for the separation of acidic compounds.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty