Would you like to log in again?
Purospher® STAR HPLC Columns and Cartridges, MilliporeSigma
Supplier: MilliporeSigma
Purospher™ Purospher® STAR, Hibar® Purospher® STAR
1.50626.0001 1.50358.1003 1.50032.0001 1.50627.0001 1.50245.0001 1.50623.0001 1.50240.0001 1.50624.0001 1.50256.0001 1.50231.0001 1.50035.0001 1.50267.0001 1.50248.0001 1.50358.0001 1.51455.0001 1.50225.0001 1.50252.0001 1.50032.0001 1.50274.0001 1.50625.0001 1.50036.0001 1.50253.0001 1.50250.0001 1.50626.0001 1.50268.0001 1.50251.0001 1.50357.0001 1.50254.0001 1.50270.0001 1.50359.0001 1.51915.0001 1.50272.0001 1.50617.1003 1.50038.0001 1.50037.0001 1.50255.0001 1.51014.0001 1.50359.1003 1.51919.1003 1.50627.0001 1.50240.0001 1.50244.0001 1.51918.0001 1.51912.0001 1.50356.0001 1.50251.1003 1.50239.0001 1.50358.0001 1.50274.0001 1.50242.0001 1.50245.0001 1.51919.0001 1.50225.0001 1.50253.0001 1.51456.0001 1.50272.0001 1.50033.0001 1.50356.0001 1.51917.0001 1.50269.0001 1.50247.0001 1.50625.0001 1.50357.0001 1.50251.0001 1.51913.0001 1.50249.0001 1.50750.0001 1.50253.0001 1.50249.0001 1.50244.0001 1.51460.0001 1.50271.0001 1.50031.0001 1.50037.0001 1.51453.0001 1.51918.1003 1.50250.0001 1.50241.0001 1.51454.0001 1.51453.0001 1.51454.0001 1.50359.0001 1.50231.0001 1.50035.0001 1.51914.0001 1.50038.0001 1.50247.0001 1.50630.0001 1.50255.0001 1.51460.0001 1.50037.1003 1.51455.1003 1.50242.0001 1.50623.0001 1.51455.0001 1.50629.0001 1.50254.0001 1.50624.0001 1.50248.0001 1.51456.1003 1.51911.0001 1.50648.1003 1.50252.0001 1.50252.1003 1.50241.0001 1.50253.0001 1.50036.1003 1.51013.0001 1.50268.0001 1.51918.0001 1.50256.0001 1.50267.0001 1.51919.0001 1.51916.0001 1.50033.0001 1.50036.0001 1.50031.0001
48219-784EA 710.68 USD InStock
CA1.50253.0001 CA48219-796 48219-784 48219-787 CA48219-793 CA48219-794 CA48219-790 48219-782 CA48219-825 10811-986 48219-814 48219-817 10811-984 CA48219-821 CA48219-789 48219-810 CA48219-787 48219-812 EM1.50626.0001 CA1.50254.0001 CA48219-784 CA48219-782 48219-770 EM1.50625.0001 48219-772 48219-803 CA48219-812 48219-806 48219-805 CA48219-810 48219-800 48219-768 10811-998 10143-996 10143-994 48219-808 CA48219-817 48219-480 48219-482 48219-488 CA1.50256.0001 CA1.50624.0001 CA1.51919.0001 CA48219-772 48219-484 CA48219-770 48219-486 48219-760 CA48219-803 CA48219-768 CA48219-766 CA48219-800 CA1.50625.0001 10812-184 10812-186 CA48219-808 CA48219-805 10812-188 CA48219-806 CA48219-480 EM1.50253.0001 48219-470 CA48219-488 48219-476 48219-754 CA48219-760 CA48219-486 48219-478 48219-472 CA48219-484 48219-474 48219-870 CA48219-482 10811-930 48219-869 EM1.50624.0001 EM1.50627.0001 CA48219-478 CA48219-754 10811-818 10811-936 CA48219-476 CA1.50627.0001 CA48219-870 CA48219-474 CA48219-472 CA48219-470 10811-820 10811-824 EM1.50750.0001 10811-826 10143-548 10811-952 10811-950 71001-970 10811-956 10143-550 10811-954 10811-838 10811-850 10811-842 10811-840 10811-846 EM1.50623.0001 10811-844 10811-848 CA1.50623.0001 CA1.50626.0001 48219-796 48219-798 48219-794 48219-825 48219-828 48219-789 48219-821 CA48219-798 CA1.51918.0001
Purospher® STAR HPLC Columns and Cartridges, MilliporeSigma
Chromatography Columns
Columns are for use with basic, neutral, metal, chelating, or any other sample format, and are ideal for the separation of difficult hydrophobic samples.
Purospher™ STAR columns are designed to provide optimum results even when loaded with large or small sample volumes and are also available in a narrow-bore format (1 mm). The wide pH range provides the required pH stability for 99% of the common analysis applications. These columns provide the power to separate even the most difficult of compound mixtures. With up to 150000 plates per meter for the 3 µm particle size and 100000 plates per meter for the 5 µm particle size, Purospher™ STAR columns ensure good baseline separations. Special derivatization and endcapping processes ensure long-term chemical stability, excellent retention stability, and long column lifetime. All Purospher™ STAR columns are guaranteed to provide absolutely reproducible results.
Ordering information: All LiChroCART® cartridges require end fittings. manuCART® and LiChroCART® series end fittings and holders are available separately. Hibar® columns include end fittings. If 4×4 mm guard columns are to be used on 4 or 4.6 mm Int.Ø. Hibar® Purospher™ STAR columns, the top fitting must be replaced with guard column holder designed to accommodate this size guard column. Other larger Int.Ø. Purospher™ STAR columns include end fittings as indicated in their descriptions. Guard columns for these larger Int.Ø columns are also available, contact your Avantor sales representative to order.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty