Would you like to log in again?
Bev-L-Edge® Micro Slides, Propper Manufacturing
Supplier: Propper
15300100 15400100
48300-364CS 284.28 USD InStock
48300-364 48300-400
Bev-L-Edge® Micro Slides, Propper Manufacturing
Slides Microscope Slides
Bev-L-Edge® microscope slides are made of first quality water-white glass and are excellent for use with blood smears or in routine microscopy.
  • Top quality ready-to-use slides for use in routine microscopy
  • Features smoothly ground beveled edges
  • Single side frosted or double side Twin-Frosted
  • Available in thickness 0.9 mm - 1.0 mm
Bev-L-Edge® Microscope Slides are made of top quality, non-corrosive, non-fogging water-white glass. Slides are priced for single use, but durable enough for repeated applications. Bev-L-Edge® Microscope Slides are available in plain or frosted (Twin-Frost). Bev-L-Edge® Microscope Slides are pre-cleaned and are perfect for use in blood smears and all routine microscopy.
Certifications: ISO 8037-1:1986; ISO 8037-2:1997
Ordering information: Packaged 1/2 gross per box (two boxes per gross).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty