Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
General Frosted and Plain Microscope Slides, Epredia™
Supplier: Epredia
2957F-601640 2957WFT-006 2957WFT-006 500-001T 510D-001T 2956-001
48303-002CS 396.03 USD InStock
48303-002 89137-981 28351-101 89137-983 CA48393-126 48382-180
General Frosted and Plain Microscope Slides, Epredia™
Slides Microscope Slides
Ideal for use in nonsterile cytology and pathology applications
  • Manufactured to the highest standards
  • Pre-cleaned for use in hematology, microbiology, urinalysis, and general laboratory applications
  • Finely ground edges
  • Frosted tab versions for manual labeling or application of printed labels
  • Plain versions are completely plain without frosted tab
  • Ready to use
Made of water-white, noncorrosive glass for maximum clarity.
Ordering information: Packaged ½ gross per pack (two packs per gross).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty