Would you like to log in again?
Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Supplier: Thermo Scientific
Colorfrost®
7951PLUS-006 6951PLUS-006 8951PLUS-006 5951PLUS-006
48312-903CS 1602.02 USD InStock
48312-903 48312-905 48312-906 48312-909
Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Slides Microscope Slides
A permanent positive charge applied to the slide surface allows slides to hold tissue sections without the need for expensive or time-consuming preparatory steps in the laboratory
This treatment virtually eliminates tissue loss during staining and electrostatically attracts frozen tissue sections and cytology preparations, binding them to the slide. Color-coded for quick and simple identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty