Would you like to log in again?
Boerner MicroTest Slide. Cs12, Wheaton
Supplier: DWK Life Sciences
798550
48351-005CS 439.95 USD InStock
48351-005
Boerner MicroTest Slide. Cs12, Wheaton
Slides Microscopy Control Slides
The WHEATON Boerner Micro Test Slide is ideal for the Boerner-Jones-Lukens microflocculation test
The slide is used for precedures involving 0.01 to 0.15mL of fluid. The slide has 10 cells arranged in 2 rows of 5 each. Each cell is 2.5mm deapp and is designed to produce flattned drops of uniform depth. The slide is manufactured from soda-lime glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty