Would you like to log in again?
Plain Micro Slides, Erie Scientific (Thermo Scientific)
Supplier: Thermo Scientific
2950WX-003
48382-142CS 1824.82 USD InStock
48382-142
Plain Micro Slides, Erie Scientific (Thermo Scientific)
Slides Microscope Slides
These Erie Scientific special white glass slides have low iron content, no green tint, and feature ground and polished edges for safe handling. Individually precleaned.
  • Corrosion-resistant
  • Conform to federal specification NNN-S-450
Ordering information: Packaged ¹/₂ gross per box, 2 boxes per gross.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty