Would you like to log in again?
Microarray Slides, Erie Scientific (Thermo Scientific)
Supplier: Thermo Scientific
C63-5876-M20 C52-5589-M20 C22-5128-M20 C50-5588-M20 C60-5590-M20 C40-5257-M20 C09-5076-M20 C18-5131-M20 C23-5133-M20
16001-348PK 426.9 USD InStock
16001-348 48382-212 48382-232 48382-220 48382-216 48382-226 48382-214 48382-208 48382-228
Microarray Slides, Erie Scientific (Thermo Scientific)
Slides Microarray Slides
These standard 25x75mm (1x3") slides feature the finest substrates available combined with the latest chemistries and surface treatments to produce superior microarrays
The SuperChip® slides are coated substrates designed for microarrays with nucleic acids as probes. Each slide features a consistent, superior surface coating to provide a uniform DNA immobilizing surface for an optimal amount of probe while minimizing nonspecific binding of labeled targets. This assures high density of reactive groups and eliminates virtually all surface artifacts. The amino silane slides are designed for microarrays fabricated with PCR products as probes.
UltraClean™ slides feature a pristine glass surface that can be used in any coating process compatible with glass such as silanization and polylysine. The slides provide a high quality “ultraclean” economical substrate for custom surface coating.
Slides with enhanced surface (ES) feature a tighter surface morphology of the glass, which results in a more uniform spot size. The ES surface may be coated with any of the attachment chemistries offered. Because the ES is microscopic it will not affect the function of microarray spotters or scanners. All slides are prepackaged in a secure, no-contact polypropylene mBox® slide handling tray, then vacuum-sealed in a foil barrier bag prior to shipping. The mBox® unit is manufactured to prevent slide contamination and withstands boiling water without softening. It contains a slide rack optimally designed for easy slide removal and allows immersion of up to 20 slides in liquids during blocking, denaturating, and post-arraying. The mBox® tray fits a standard microtiter plate format for centrifuging. The mTub™ disposable microarray tray is designed to fit the mBox® tray. The mTub™ disposable microarray tray will accept most glass and stainless steel boats.
All slides are manufactured from the highest-quality low fluorescence glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty