Would you like to log in again?
KIMAX® Babcock Bottle, Milk Test, 8%, Sealed, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
1003S-8
48711-056CS 447.02 USD InStock
48711-056
KIMAX® Babcock Bottle, Milk Test, 8%, Sealed, Kimble Chase
Bottles Dairy Testing Bottles
Designed for use in AOAC®/APHA test procedures.
Neck is graduated to 8% in 0.1% subdivisions. Etched vertical line, used to anchor calipers is centered in a contrasting darkened band which makes visualization easier. Marked with 'Sealed 3' for use in states requiring this marking.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty