Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMAX® Babcock Bottle, Ice Cream Test, 10%, Kimble Chase

Supplier: DWK Life Sciences
KIMAX®
1025-10
48711-100CS 393.62 USD InStock
48711-100
KIMAX® Babcock Bottle, Ice Cream Test, 10%, Kimble Chase
Bottles Dairy Testing Bottles
Designed for testing butter fat content.

Neck is graduated to 10% in 0.1% subdivisions. Scale is permanent brown stain.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty