Would you like to log in again?
KIMAX® Babcock Bottle, Cream and Cheese Test, 50%, Sealed, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
2015S-50
48712-007CS 340.92 USD InStock
48712-007
KIMAX® Babcock Bottle, Cream and Cheese Test, 50%, Sealed, Kimble Chase
Bottles Dairy Testing Bottles
Designed for testing butter fat content.
  • Scale is permanent brown stain
Neck is graduated to 50% in 0.5% subdivisions. Marked 'Sealed 3' for use in states requiring this special marking.
Certifications: Made in accordance with the specifications of the American Dairy Science Association, and the AOAC®/APHA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty