Would you like to log in again?
Paley Bottle, Cheese Test, 50%, Kimble Chase
Supplier: DWK Life Sciences
509-50
48719-000CS 347.93 USD InStock
48719-000
Paley Bottle, Cheese Test, 50%, Kimble Chase
Bottles Dairy Testing Bottles
Facilitates the introduction of solid or viscous materials directly into the bottle, which can then be weighed.
Neck is graduated to 50% in 0.5% subdivisions. The bottle is designed so that it can be used in regular test bottle centrifuge equipment.
Ordering information: Supplied with directions and three styrene-butadiene stoppers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty