Would you like to log in again?
KIMAX® Paley Bottle, 20%, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
508-20
48719-021CS 324.99 USD InStock
48719-021
KIMAX® Paley Bottle, 20%, Kimble Chase
Bottles Dairy Testing Bottles
Designed for use in APHA 15.8 test procedures. Developed to facilitate the introduction of solid or viscous materials directly into the bottle.
  • Calibrated for a 9 gram sample
  • Scale is permanent brown stain
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Ordering information: Three rubber stoppers are supplied with each bottle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty