Would you like to log in again?
KIMAX® Babcock Bottle, Ice Cream and Half and Half Tests, 20%, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
516-20
48720-009CS 468.96 USD InStock
48720-009
KIMAX® Babcock Bottle, Ice Cream and Half and Half Tests, 20%, Kimble Chase
Bottles Dairy Testing Bottles
KIMAX® bottle is filled through the graduated neck.
Scale is permanent brown stain. Designed for use in APHA test procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty