Would you like to log in again?
MILL UNITIZED JAR 2-TIER
Supplier: U S STONEWARE CORP
Jar Mills^ Space-saving roller mills grind algae, crystalline chemicals, coal, inks, paints, pigments, plant aggregates, rocks, soils. Tiers: 2; Maximum Weight/Tier: 60 lb. Roll Length: 13. Dimensions (LxWxH): 20 1/4"x12 1/8"x34 3/8". Motor HP: 1/4.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MILL UNITIZED JAR 2-TIER 48902-006 M93122DC-1 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty