Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sample Wrapper, Thomas Scientific
Supplier: Thomas Scientific
6514F70
48956-142PK 170.8 USD InStock
48956-142
Sample Wrapper, Thomas Scientific
Seals General Purpose Sealing Films
Sample Wrapper absorbs heat quickly for infrared ignition; die-cut from unsized paper
Sample area 32 x 30 mm, with 8 x 38 mm extending strip as fuse for ignition.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty