Would you like to log in again?
J.T.Baker® Robotic Tips Type ST
Supplier: Ritter, part of Avantor
J.T.Baker®
49001-11061 49001-00001 49000-11061 49001-01061 49000-00001 49000-00061
49001-0106CS 518.23 USD InStock
49001-0106 49000-0000 49001-0000 49000-1106 49000-0006 49001-1106
J.T.Baker® Robotic Tips Type ST
Pipette Tips
Suitable for use on Euroimmun Analyzer. Available in conductive version, with or without filter.
  • Manufactured under cleanroom conditions (cleanroom class 8)
  • Conductive tips for liquid level sensing
  • Ideal for dosing even small volumes with high precision and without contamination
J.T.Baker® robotic tips type ST are suitable for use on Euroimmun Analyzer and other robotic instruments using the same pipetting head.
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty