Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MLA D-Tipper Pipettors, VistaLab
Supplier: VistaLab
MLA
1055C 1148C 1132 1051PK 1147C 1053C 1057C 1152C 1149C 1151C 1146C 1054C 1160C 1056PK 1154C 1058C 1143C 1163C 1052PK 1060C 1159C 1141C 1055PK 1059PK 1052C 1153C 1157C 1050-0999 1142C 1057PK 1054PK 1150C 1050-0998 1162C 1056CLT 1060PK 1056C 1059C 1051C 1144C
49000-536EA 427.8 USD InStock
49000-536 53514-036 75814-370 75814-392 53514-038 49000-534 75814-390 49000-538 89368-672 89368-674 53514-040 53514-042 53514-086 53514-088 75814-388 53514-044 53514-100 53514-046 89368-670 53514-026 49000-546 49000-544 49000-542 89368-662 75813-772 89368-664 53514-090 89368-666 89368-668 53514-092 53514-094 49000-540 53514-030 53514-096 53514-076 53514-032 53514-098 53514-034 89368-660 75814-374
MLA D-Tipper Pipettors, VistaLab
Pipettes Repetitive Pipettors
MLA pipettors feature a built-in tip ejector and single-stroke 'to deliver' operation.
  • Glove guard
  • Available in a wide range of fixed volumes
  • Light plunger forces for easy operation with thumb/forefinger
  • Built-in tip ejector
  • Individually serialized
  • Lifetime warranty on parts
MLA pipettes are made of precision-engineered aluminum alloy and stainless steel.
These pipettors are available in 30 fixed volumes ranging from 5 to 1000 μl, or as a selectable model that is compatible at volumes of 50, 100 and 200 μl. Pipettors feature color-coded bodies for easy volume identification.
They are designed for highly accurate delivery and proven performance.
Accessories information: Seal Kits for MLA Pipettors are available separately. Seal kits contain lubricant, seals, and instructions.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty