Would you like to log in again?
GeneMate Mini Dry Bath with Heated Lid
Supplier: Benchmark Scientific
BSH200-HL-GM
490000-072EA 1033.52 USD InStock
490000-072
GeneMate Mini Dry Bath with Heated Lid
Block Heaters Digital Block Heaters
This Mini Dry Bath with Heated Lid is the first mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation.
  • 2-year full replacement warranty
  • Heated lid prevents evaporation & condensation
  • Compact, fits in the palm of your hand
  • Independent temperature control of lid & block
  • Exchangeable blocks for 0.2 to 0.5 mL tubes
  • Room temperature to 37°C in just 2 minutes
  • Blocks Sold Separately
Both the block and lid temperature can be selected independently in increments of 0.1°C, making this the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions & other temperature hyper-sensitive procedures.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty