Would you like to log in again?
Polycarbonate Boxes, Crystal Industries
Supplier: Crystal
PB2-81 PB2-100
490000-792EA 47.88 USD InStock
490000-792 490000-790
Polycarbonate Boxes, Crystal Industries
Boxes Cryoboxes
Polycarbonate 81 or 100 cell 2" boxes hold most popular cryo and cold storage vials and fit into standard freezer racks.
  • Boxes have clear lids with locator grids to facilitate and sample identification
  • Polycarbonate boxes are designed to withstand temperature extremes of –196 to 121 °C
Polycarbonate boxes have drain holes for venting and safety and the keyed lid prevents misalignment of grid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty