Would you like to log in again?
GeneMate LowMelt Agarose
Supplier: GeneMate
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LM-25G LM-125G
490001-596EA 1547.71 USD InStock
490001-596 490001-598
GeneMate LowMelt Agarose
Electrophoresis Gels Agarose and Acrylamide
  • Electrophoresis of DNA Fragments, 1000 bp
  • Tissue and Cell Culture
Low melting temperature agaroses result from a derivatization process by organic synthesis, which generates methoxylate groups from the basic agarose structure. This results in agaroses with low melting and gelling temperatures when compared to standard agaroses. Why use a low melting point agarose? The low melting temperature allows for the recovery of nucleic acids below their denaturing temperature, thus preventing damage to the nucleic acid. Low gelling temperature ensures the agarose will be in a liquid state at a temperature range where in-gel manipulations can be performed without prior extraction of the DNA from the gel slice. GeneMate LowMelt has the highest melting and gelling temperatures in this class of low melt agaroses, as well as the highest gel strength.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty