Would you like to log in again?
GeneMate LowMelt GQA Agarose
Supplier: GeneMate
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LM-GQT-125G LM-GQT-25G
490001-600EA 1260.6 USD InStock
490001-600 490001-602
GeneMate LowMelt GQA Agarose
Electrophoresis Gels Agarose and Acrylamide
GeneMate LowMelt GQA is a low melting temperature agarose with the highest resolving capacity for large DNA fragments >1000bp, including PCR products. This agarose is Genetic Quality Analyzed (GQA) and carries that certification. This ensures that in-gel applications can be performed in remelted agarose, avoiding difficult DNA extraction steps. GeneMate LowMelt GQA agarose is ideal for digestion by agarose enzymes, making it very easy to recover large DNA fragments suitable for cloning or enzymatic processing.
  • Genetic Quality Tested
  • DNA/RNA Separation, 1Kb
  • Anaysis and Recovery of DNA Fragments
  • In-Gel Enzymatic processing, Digestion, Ligation, PCR
  • Preparative Electrophoresis
  • DNA Binding: None detected
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty