Would you like to log in again?
GeneMate Sieve GQA Low Melt Agarose
Supplier: GeneMate
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LM-SIEVE-125G LM-SIEVE-500G LM-SIEVE-25G
490001-604EA 896.41 USD InStock
490001-604 490001-608 490001-606
GeneMate Sieve GQA Low Melt Agarose
Electrophoresis Gels Agarose and Acrylamide
  • DNA/RNA Separation, 1 Kb
  • Analytical and Preparative Electrophoresis
  • In-gel enzymatic processing, Digestion, Ligation, PCR
  • Preparative Electrophoresis
GeneMate Sieve GQA Agarose is a low melting temperature agarose with the highest resolving capacity for DNA fragments smaller than 1000 bp, especially PCR products ranging from 200 to 800 bp. This agarose is Genetic Quality Analyzed, GQA, and carries that certification. This ensures that in-gel applications can be performed in remelted agarose, avoiding difficult DNA extraction steps. GeneMate Sieve GQA Agarose is ideal for digestion by agarose enzymes, making it very easy to recover large DNA fragments suitable for cloning or enzymatic processing. It can also be used at high concentrations, forming gels with excellent clarity and a higher sieving capacity than standard melting agaroses. GeneMate Sieve GQA gels are very easy to handle, even at concentrations as low as 3%. Genetic Quality Testing Certified.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty