Would you like to log in again?
GeneMate 3:1 Agarose
Supplier: GeneMate
MS-12-125G MS-12-25G
490001-610EA 792.93 USD InStock
490001-610 490001-612
GeneMate 3:1 Agarose
Electrophoresis Gels Agarose and Acrylamide
GeneMate 3:1 Agarose is a molecular screening agarose designed to have a larger gel network than GeneMate Hi-Res and is recommended for use in separating DNA fragments smaller than 1000 bp
  • Very low DNA binding
Like all GeneMate agaroses, GeneMate 3:1 delivers higher gel strength. At the same time, the gel is extremely firm but still flexible when handled, minimizing the danger of cracking or breaking. It has the same melting and gelling temperatures as standard agaroses, allowing faster and easier preparation of gels. GeneMate 3:1 is recommended for all analytical applications and can be used when DNA is recovered for subsequent use in enzymatic procedures. Excellent for blotting and for resolution of small DNA fragments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty