Would you like to log in again?
8- Strip Ultraflux PCR Tubes and Caps, Scientific Specialties
Supplier: Scientific Specialties
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3101-02-200IS 3131-00-210IS 3101-09-200IS 3102-00-200IS 3111-09-210IS 3131-09-210IS 3111-00-210IS
490003-606BX 273.9 USD InStock
490003-606 490003-538 490004-430 490003-530 490003-574 490003-540 490003-562
8- Strip Ultraflux PCR Tubes and Caps, Scientific Specialties
Tubes PCR Tubes
Ultraflux PCR tubes are designed to precisely fit most thermal cyclers.
  • Convenient tube and cap combination packs
  • Secure evaporation-resistant seal between cap lid and tube
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Tubes are pre-numbered for easy well identification
  • Crystal-clear clarity and easy to visualize content of each well
  • Ideal for enhancing signal strength for qPCR assays
Certified free DNase, RNase, and detectable DNA.
Ordering information: GeneMate 96-Well PCR tube storage racks are available separately.
Packaging: Tubes and caps are packed equally in one pack.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty