Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GeneMate SnapStrip® PCR Tubes with Individual Attached Dome Caps, 8-Strip 0.2 ml, Scientific Specialties

Supplier: Scientific Specialties

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

3240-09-210IS 3240-00-212IS
490003-704BX 420.51 USD InStock
490003-704 490003-692
GeneMate SnapStrip® PCR Tubes with Individual Attached Dome Caps, 8-Strip 0.2 ml, Scientific Specialties
Tubes PCR Tubes
The SnapStrip is an attached 8-strip of tubes with each tube in the strip having its own individual attached dome cap. Another unique and innovative feature of the tubes are user's ability to create a strip of less than 8 tubes with caps by simply breaking the strip where desired.

  • Ulta-thin wall for optimum heat transfer
  • Certified free from DNase, RNase, and detectable DNA

A strip of 2 or more tubes is easier and safer to use and manipulate than are single tubes with caps.

Flexibility: create a 4-strip today, create a 6-strip tomorrow. The individual tubes may also be used with the 0.2 or 0.5 ml adapters.

Packaging: Packaged as 120 Strips of 8, case has 10 packs of 120 strips of 8.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty