Would you like to log in again?
GeneMate SnapStrip® 8-Strip Standard PCR Tubes with Individually Attached Flat Caps, Scientific Specialties
Supplier: Scientific Specialties
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3245-00-210IS 3246-00-210IS
490003-708BX 194.94 USD InStock
490003-708 490003-706
GeneMate SnapStrip® 8-Strip Standard PCR Tubes with Individually Attached Flat Caps, Scientific Specialties
Tubes PCR Tubes
Designed to precisely fit most thermal cyclers. Ideal for qPCR (real-time PCR) and standard PCR. Secure evaporation-resistant seal between cap lid and tube. Individual tubes have caps closing at an angle.
  • Angled caps prevent interference from adjacent tubes in thermal cycler
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Easy to break-a-part tubes
  • Optically-clear flat caps
  • Crystal-clear clarity, easy to visualize content of each well
  • Available in full range of colors or white (enhances qPCR signals)
  • Certified free DNase, RNase, and detectable DNA
Real-Time PCR strip tubes combine a flat cap with high clarity manufacturing. The high clarity means it's ideally suited for real-time PCR (QPCR) applications where excitation and emission takes through the cap. The flat cap aids transmission and also makes an ideal writing or marking surface. These strip tubes fit a diverse range of thermal cyclers. Two tube profiles, standard and low-profile, ensure universal compatibility.
Ordering information: GeneMate 96-well PCR tube storage racks are available separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty