Would you like to log in again?
GeneMate UltraFlux® I, 8-Strip Standard PCR Tubes with Individually Attached, Optically-Clear Flat Caps, Scientific Specialties
Supplier: Scientific Specialties
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3247-00-210IS 3247-09-210IS 3248-00-210IS
490003-726BX 202.61 USD InStock
490003-726 490003-722 490003-710
GeneMate UltraFlux® I, 8-Strip Standard PCR Tubes with Individually Attached, Optically-Clear Flat Caps, Scientific Specialties
Tubes PCR Tubes
Designed to precisely fit most thermal cyclers. Ideal for qPCR (real-time PCR) and standard PCR. Secure evaporation-resistant seal between cap lid and tube. Individual tubes have caps closing at an angle.
  • Angled caps prevent interference from adjacent tubes in thermal cycler
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Robust interwell linkages makes tubes more rigid and sturdy
  • Optically-clear flat caps
  • Crystal-clear clarity, easy to visualize content of each well
Available in full range of colors or white (enhances qPCR signals).
Certified free DNase, RNase, and detectable DNA.
Ordering information: GeneMate 96-well PCR tube storage racks are available separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty