Would you like to log in again?
GeneMate Stacking 96-Well PCR Work Rack, Scientific Specialties
Supplier: Scientific Specialties
5240-09-22IS1
490003-980BX 153.51 USD InStock
490003-980
GeneMate Stacking 96-Well PCR Work Rack, Scientific Specialties
Racks Tube Racks
The latest design has a positive lid locator meaning the lid will not become easily dislodged. Yet only a gentle squeeze on each side is all that is needed to unlock the lid. The racks are manufactured in five distinctive colors: Blue, Green, Yellow, Violet, and Red.
  • Easy stacking with or without lid
  • Lid clicks into place
  • Accommodates PCR plates, strips, & tubes
  • Ideal for bench, refrigerator, or freezer
  • Rack dimensions with lid in place: 1.54" x 3.86" x 5.43" (39 x 98 x 138 mm)
  • Rack dimensions with lid removed: 1.18" x 3.86" x 5.43" (30 x 98 x 138 mm)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty