Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tube Storage Boxes, 15 and 50 ml, GeneMate

Supplier: GeneMate
5570-02-68IS1 5580-01-68IS1
490004-202BX 92.89 USD InStock
490004-202 490004-200
Tube Storage Boxes, 15 and 50 ml, GeneMate
Boxes Cryoboxes
These polypropylene tube storage boxes are ideal for freezer storage.

  • Clear lid for easy location of tubes
  • Removable base for use in ice bath
  • Dimensions: 5.38×5.38×5.00" with lid
  • Frosted writing surface on two opposite lid sides for easy labeling
  • Green box holds 36 tubes
  • Blue box holds 16 tubes
  • White numerical indexing
  • 2 Racks/Pack

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty